Gene Transcripts

Gene Symbol
Olfr807
Entrez Gene ID
258931
Gene Name
olfactory receptor 807
Gene Aliases
MOR117-1
Chromosome Location
Chr. 10: 129754495 - 129757800 on Build GRCm38
UniGene ID
Mm.223330
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146929.1 NP_667140.1 1 630
GenBank mRNA BC138074.1 - 1 667

View Full Product Details