Gene Transcripts

Gene Symbol
Olfr392
Entrez Gene ID
259008
Gene Name
olfactory receptor 392
Gene Aliases
MOR135-9
Chromosome Location
Chr. 11: 73814124 - 73816877 on Build GRCm38
UniGene ID
Mm.223395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147006.2 NP_667217.2 2 909
GenBank mRNA CB173273.1 - 1 357

View Full Product Details