Gene Transcripts
- Gene Symbol
- Pum2
- Entrez Gene ID
- 80913
- Gene Name
- pumilio RNA-binding family member 2
- Gene Aliases
- 5730503J23Rik, Pumm2, mKIAA0235
- Chromosome Location
- Chr. 12: 8674134 - 8752583 on Build GRCm38
- UniGene ID
- Mm.341243
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001160219.1 |
NP_001153691.1
|
18, 19 |
3002 |
|
NM_001160220.1 |
NP_001153692.1
|
18, 19 |
2996 |
|
NM_001160221.1 |
NP_001153693.1
|
18, 19 |
2811 |
|
NM_001160222.1 |
NP_001153694.1
|
17, 18 |
2574 |
|
NM_030723.2 |
NP_109648.2
|
18, 19 |
2926 |
|
GenBank mRNA |
AF315590.1 |
- |
18, 19 |
2942 |
|
AK048119.1 |
- |
12, 13 |
1865 |
|
AK122225.1 |
- |
19, 20 |
2844 |
|
AK134185.1 |
- |
18, 19 |
2810 |
|
AK141682.1 |
- |
18, 19 |
2858 |
|
AK147617.1 |
- |
19, 20 |
2944 |
|
AK161357.1 |
- |
7, 8 |
1167 |
|
AK163964.1 |
- |
17, 18 |
2549 |
|
AY027917.1 |
- |
18, 19 |
2842 |
|
BC013765.1 |
- |
2, 3 |
177 |
|
BC041773.1 |
- |
16, 17 |
2548 |
3986207 |
View Full Product Details