Gene Transcripts

Gene Symbol
2810428I15Rik
Entrez Gene ID
66462
Gene Name
RIKEN cDNA 2810428I15 gene
Gene Aliases
Chromosome Location
Chr. 8: 70504295 - 70506773 on Build GRCm38
UniGene ID
Mm.28242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025577.2 NP_079853.1 2 124
GenBank mRNA AK013181.1 - 2 125
AK160511.1 - 2 68
BC028495.1 - 2 130 3370477

View Full Product Details