Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_183275.2 | NP_899098.2 | 4 | 1054 | |
GenBank mRNA | AK003325.1 | - | 4 | 1031 | |
AK146639.1 | - | 4 | 1055 | ||
BC019666.1 | - | 1 | 368 | ||
BC094625.1 | - | 2 | 488 | 1515023 | |
BC132029.1 | - | 3 | 694 | ||
BE949440.1 | - | 1 | 245 |