Gene Transcripts

Gene Symbol
Uqcc3
Entrez Gene ID
107197
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 3
Gene Aliases
AI462493, Cbp4
Chromosome Location
Chr. 19: 8880014 - 8880933 on Build GRCm38
UniGene ID
Mm.381181
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160356.1 NP_001153828.1 1 168
GenBank mRNA AK131602.1 - 1 159
AK131892.1 - 1 132
AK132116.1 - 1 159
AY941793.1 - 1 159
BC029863.1 - 1 93 4460168

View Full Product Details