Gene Transcripts

Gene Symbol
Aif1l
Entrez Gene ID
108897
Gene Name
allograft inflammatory factor 1-like
Gene Aliases
2810003C17Rik, AI043124, C87647, Iba2
Chromosome Location
Chr. 2: 31950303 - 31973442 on Build GRCm38
UniGene ID
Mm.24838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145144.1 NP_660126.1 4, 5 249
GenBank mRNA AB035322.1 - 4, 5 201
AK028955.1 - 4, 5 207
AK045539.1 - 4, 5 279
BC024599.1 - 4, 5 249 5044495

View Full Product Details