Gene Transcripts

Gene Symbol
Bad
Entrez Gene ID
12015
Gene Name
BCL2-associated agonist of cell death
Gene Aliases
AI325008, Bbc2
Chromosome Location
Chr. 19: 6941861 - 6951905 on Build GRCm38
UniGene ID
Mm.4387
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285453.1 NP_001272382.1 2 108
NM_007522.3 NP_031548.1 2 625
GenBank mRNA AK131666.1 - 2 70
AK151792.1 - 2 73
AK169002.1 - 2 70
BC006762.1 - 2 593 3257589
L37296.1 - 2 633

View Full Product Details