Gene Transcripts

Gene Symbol
Lbp
Entrez Gene ID
16803
Gene Name
lipopolysaccharide binding protein
Gene Aliases
Bpifd2, Ly88
Chromosome Location
Chr. 2: 158306493 - 158332852 on Build GRCm38
UniGene ID
Mm.218846
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008489.2 NP_032515.2 14, 15 1577
GenBank mRNA BC004795.1 - 14, 15 1465
BQ176753.1 - 1, 2 424
X99347.1 - 14, 15 1527

View Full Product Details