Gene Transcripts

Gene Symbol
Mbd3
Entrez Gene ID
17192
Gene Name
methyl-CpG binding domain protein 3
Gene Aliases
AI181826, AU019209
Chromosome Location
Chr. 10: 80392539 - 80400702 on Build GRCm38
UniGene ID
Mm.7142
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF072248.1 - 4 701
BC038264.1 - 4 502 5250237
BQ042794.1 - 3 269
CK625083.1 - 4 521

View Full Product Details