Gene Transcripts

Gene Symbol
Them6
Entrez Gene ID
223626
Gene Name
thioesterase superfamily member 6
Gene Aliases
4930572J05Rik
Chromosome Location
Chr. 15: 74721234 - 74724373 on Build GRCm38
UniGene ID
Mm.268603
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198607.1 NP_941009.1 1 404
GenBank mRNA AK019809.1 - 1 15
BC047266.1 - 1 404 6307114

View Full Product Details