Gene Transcripts

Gene Symbol
Tmem191c
Entrez Gene ID
224019
Gene Name
transmembrane protein 191C
Gene Aliases
4933405M22Rik, D16Bwg1494e, MNCb-4137
Chromosome Location
Chr. 16: 17276300 - 17278661 on Build GRCm38
UniGene ID
Mm.246388
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177473.3 NP_803424.2 10 876
GenBank mRNA AB041601.1 - 7 1193
AK016675.1 - 8 1075
BC058760.1 - 6 971
BC098238.1 - 6 503

View Full Product Details