Gene Transcripts
- Gene Symbol
- Nkx2-4
- Entrez Gene ID
- 228731
- Gene Name
- NK2 homeobox 4
- Gene Aliases
- 1700001P03Rik, Nkx-2.4, tinman
- Chromosome Location
- Chr. 2: 147083337 - 147085439 on Build GRCm38
- UniGene ID
- Mm.315498
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023504.1 |
NP_075993.1
|
1 |
65 |
|
View Full Product Details