Gene Transcripts

Gene Symbol
Neurl1b
Entrez Gene ID
240055
Gene Name
neuralized homolog 1b (Drosophila)
Gene Aliases
C230078M08Rik, EG240055
Chromosome Location
Chr. 17: 26414965 - 26446349 on Build GRCm38
UniGene ID
Mm.133211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081656.2 NP_001075125.1 5 1616
GenBank mRNA DQ839448.1 - 5 1616
GQ414760.1 - 5 1616

View Full Product Details