Gene Transcripts

Gene Symbol
Olfr1225
Entrez Gene ID
258893
Gene Name
olfactory receptor 1225
Gene Aliases
MOR233-11
Chromosome Location
Chr. 2: 89170219 - 89171245 on Build GRCm38
UniGene ID
Mm.377737
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146891.2 NP_667102.2 1 853
GenBank mRNA BC145849.1 - 1 853

View Full Product Details