Gene Transcripts

Gene Symbol
Prss33
Entrez Gene ID
353130
Gene Name
protease, serine 33
Gene Aliases
Eos, mT6, tryptase-6
Chromosome Location
Chr. 17: 23833338 - 23836126 on Build GRCm38
UniGene ID
Mm.261689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BQ893522.1 - 1 402

View Full Product Details