Gene Transcripts

Gene Symbol
Pram1
Entrez Gene ID
378460
Gene Name
PML-RAR alpha-regulated adaptor molecule 1
Gene Aliases
AY665714
Chromosome Location
Chr. 17: 33638056 - 33645706 on Build GRCm38
UniGene ID
Mm.272606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002842.2 NP_001002842.2 2 808
GenBank mRNA AK169990.1 - 2 798
AY665714.1 - 2 724
BC141100.1 - 2 713 9055712

View Full Product Details