Gene Transcripts
- Gene Symbol
- Mrgpra5
- Entrez Gene ID
- 404235
- Gene Name
- MAS-related GPR, member A5
- Gene Aliases
- MrgA5
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.484774
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207534.1 |
NP_997417.1
|
|
847 |
|
View Full Product Details