Gene Transcripts

Gene Symbol
Atad2
Entrez Gene ID
70472
Gene Name
ATPase family, AAA domain containing 2
Gene Aliases
2610509G12Rik
Chromosome Location
Chr. 15: 58094047 - 58135139 on Build GRCm38
UniGene ID
Mm.221758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027435.2 NP_081711.2 13, 14 1681
GenBank mRNA AK029867.1 - 6, 7 828
AK032877.1 - 13, 14 1682

View Full Product Details