Gene Transcripts

Gene Symbol
Fance
Entrez Gene ID
72775
Gene Name
Fanconi anemia, complementation group E
Gene Aliases
2810451D06Rik, AI415634, AW209126
Chromosome Location
Chr. 17: 28313530 - 28326574 on Build GRCm38
UniGene ID
Mm.201859
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163819.1 NP_001157291.1 2 430
NM_001163820.1 NP_001157292.1 2 430
GenBank mRNA AK013325.1 - 2 394
AK135017.1 - 3 334
AK172283.1 - 2 414
BC016538.1 - 2 388
BC039987.1 - 2 393
BY740269.1 - 2 414

View Full Product Details