Gene Transcripts

Gene Symbol
Mss51
Entrez Gene ID
74843
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
4833444M15Rik, Zmynd17
Chromosome Location
Chr. 14: 20482862 - 20498131 on Build GRCm38
UniGene ID
Mm.295241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029104.1 NP_083380.1 3, 4 661
GenBank mRNA AK014794.1 - 3, 4 442
BC119773.1 - 3, 4 554 40107223
BC119816.1 - 3, 4 554 40107221

View Full Product Details