Gene Transcripts

Gene Symbol
Hrh4
Entrez Gene ID
170704
Gene Name
histamine receptor H4
Gene Aliases
Chromosome Location
Chr. 18: 4365429 - 4382599 on Build Rnor_6.0
UniGene ID
Rn.162272
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131909.1 NP_571984.1 3 688
GenBank mRNA AF358860.1 - 3 688

View Full Product Details