Gene Transcripts

Gene Symbol
Ndufaf6
Entrez Gene ID
297821
Gene Name
NADH dehydrogenase (ubiquinone) complex I, assembly factor 6
Gene Aliases
RGD1309085
Chromosome Location
Chr. 5: 24297169 - 24320804 on Build Rnor_6.0
UniGene ID
Rn.94436
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276441.1 NP_001263370.1 6 663
GenBank mRNA FQ215404.1 - 6 663

View Full Product Details