Gene Transcripts

Gene Symbol
Ldlr
Entrez Gene ID
300438
Gene Name
low density lipoprotein receptor
Gene Aliases
LDLRA
Chromosome Location
Chr. 8: 22750425 - 22773305 on Build Rnor_6.0
UniGene ID
Rn.10483
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175762.2 NP_786938.1 5 889
GenBank mRNA X13722.1 - 5 889

View Full Product Details