Gene Transcripts

Gene Symbol
Btn1a1
Entrez Gene ID
306956
Gene Name
butyrophilin, subfamily 1, member A1
Gene Aliases
Chromosome Location
Chr. 17: 43899565 - 43908986 on Build Rnor_6.0
UniGene ID
Rn.163350
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170337.1 NP_001163808.1 4 785
XM_006253970.2 XP_006254032.1 5 1164
GenBank mRNA AY327120.1 - 4 789

View Full Product Details