Gene Transcripts

Gene Symbol
Cpne5
Entrez Gene ID
309650
Gene Name
copine V
Gene Aliases
Chromosome Location
Chr. 20: 6419033 - 6500523 on Build Rnor_6.0
UniGene ID
Rn.21919
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107616.1 NP_001101086.1 12 935
GenBank mRNA AM747282.1 - 12 837

View Full Product Details