Gene Transcripts

Gene Symbol
Pus7l
Entrez Gene ID
315264
Gene Name
pseudouridylate synthase 7 homolog (S. cerevisiae)-like
Gene Aliases
RGD1304692
Chromosome Location
Chr. 7: 135765307 - 135803586 on Build Rnor_6.0
UniGene ID
Rn.41818
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729847.4 XP_002729893.2 2 649
XM_006242271.2 XP_006242333.1 2 956

View Full Product Details