Gene Transcripts

Gene Symbol
Olr289
Entrez Gene ID
405041
Gene Name
olfactory receptor 289
Gene Aliases
Chromosome Location
Chr. 1: 212836278 - 212837207 on Build Rnor_6.0
UniGene ID
Rn.141743
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000767.1 NP_001000767.1 1 768

View Full Product Details