Gene Transcripts

Gene Symbol
Padi1
Entrez Gene ID
54282
Gene Name
peptidyl arginine deiminase, type I
Gene Aliases
Pdi1
Chromosome Location
Chr. 5: 159338850 - 159371313 on Build Rnor_6.0
UniGene ID
Rn.30030
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019332.1 NP_062205.1 15, 16 1878
GenBank mRNA AB010998.1 - 15, 16 1878

View Full Product Details