Gene Transcripts

Gene Symbol
Adrm1
Entrez Gene ID
65138
Gene Name
adhesion regulating molecule 1
Gene Aliases
Gp110
Chromosome Location
Chr. 3: 175548060 - 175552943 on Build Rnor_6.0
UniGene ID
Rn.9320
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031708.1 NP_113896.1 7 875
XM_006235822.1 XP_006235884.1 7 996
GenBank mRNA AB032742.1 - 7 875
BC061773.1 - 7 891 5621873

View Full Product Details