Gene Transcripts

Gene Symbol
Gatm
Entrez Gene ID
81660
Gene Name
glycine amidinotransferase (L-arginine:glycine amidinotransferase)
Gene Aliases
Chromosome Location
Chr. 3: 114711570 - 114728155 on Build Rnor_6.0
UniGene ID
Rn.17661
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031031.2 NP_112293.1 3 503
GenBank mRNA AY625271.1 - 1 91
BC081785.1 - 3 503 7132560
U07971.1 - 3 495

View Full Product Details