Gene Transcripts

Gene Symbol
Nell1
Entrez Gene ID
81733
Gene Name
NEL-like 1 (chicken)
Gene Aliases
Chromosome Location
Chr. 1: 105349069 - 106218956 on Build Rnor_6.0
UniGene ID
Rn.10695
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031069.1 NP_112331.1 11 1159
GenBank mRNA U48246.1 - 11 1159

View Full Product Details