Gene Transcripts

Gene Symbol
Gtf2a1
Entrez Gene ID
83830
Gene Name
general transcription factor IIA, 1
Gene Aliases
Tf2a, TfIIa
Chromosome Location
Chr. 6: 115321110 - 115352659 on Build Rnor_6.0
UniGene ID
Rn.24468
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022208.1 NP_071544.1 6 544
XM_006240410.2 XP_006240472.1 6 819
GenBank mRNA AF000943.1 - 6 544

View Full Product Details