Gene Transcripts

Gene Symbol
Lcn8
Entrez Gene ID
366008
Gene Name
lipocalin 8
Gene Aliases
RGD1306747
Chromosome Location
Chr. 3: 2871890 - 2875682 on Build Rnor_6.0
UniGene ID
Rn.220344
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128183.1 NP_001121655.1 5 462
XM_006233632.2 XP_006233694.2 5 603
GenBank mRNA DQ537491.1 - 5 448

View Full Product Details