Gene Transcripts

Gene Symbol
Lhx5
Entrez Gene ID
124451
Gene Name
LIM homeobox 5
Gene Aliases
Chromosome Location
Chr. 12: 41663000 - 41671437 on Build Rnor_6.0
UniGene ID
Rn.54653
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139036.1 NP_620605.1 2 655
GenBank mRNA L35572.1 - 2 655

View Full Product Details