Gene Transcripts

Gene Symbol
Plp1
Entrez Gene ID
24943
Gene Name
proteolipid protein 1
Gene Aliases
Plp
Chromosome Location
Chr. X: 107479485 - 107511355 on Build Rnor_6.0
UniGene ID
Rn.4550
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030990.2 NP_112252.1 6 868
XM_006257293.2 XP_006257355.1 9 1150
XM_008773401.1 XP_008771623.1 9 1254
GenBank mRNA BC093596.1 - 5 729
FQ211530.1 - 6 841
FQ212519.1 - 5 477
FQ212680.1 - 5 582
FQ213540.1 - 5 582
FQ213841.1 - 5 582
FQ213943.1 - 5 582
FQ214012.1 - 5 582
FQ214050.1 - 5 582
FQ214153.1 - 5 583
FQ214556.1 - 5 582
M11185.1 - 6 843
M14126.1 - 3 196
M25888.1 - 4 408
X02809.1 - 6 772

View Full Product Details