Gene Transcripts

Gene Symbol
Clps
Entrez Gene ID
25680
Gene Name
colipase, pancreatic
Gene Aliases
COLQ
Chromosome Location
Chr. 20: 5803444 - 5806112 on Build Rnor_6.0
UniGene ID
Rn.6714
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013139.1 NP_037271.1 2 232
XM_008772711.1 XP_008770933.1 2 243
GenBank mRNA M33333.1 - 2 191
M58370.1 - 2 232

View Full Product Details