Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008217.1 | NP_001008218.1 | 4 | 519 | |
GenBank mRNA | BC127466.1 | - | 4 | 539 | |
CF110813.1 | - | 4 | 473 | ||
D30804.1 | - | 4 | 511 | ||
FQ226344.1 | - | 4 | 526 | ||
FQ231713.1 | - | 4 | 526 |