Gene Transcripts

Gene Symbol
Ildr1
Entrez Gene ID
303914
Gene Name
immunoglobulin-like domain containing receptor 1
Gene Aliases
RGD1307792
Chromosome Location
Chr. 11: 67004036 - 67037184 on Build Rnor_6.0
UniGene ID
Rn.155292
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC166709.1 - 7 1594

View Full Product Details