Gene Transcripts

Gene Symbol
Adamtsl3
Entrez Gene ID
308787
Gene Name
ADAMTS-like 3
Gene Aliases
Chromosome Location
Chr. 1: 144238958 - 144561065 on Build Rnor_6.0
UniGene ID
Rn.38679
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107533.2 NP_001101003.2 11, 12 1450
XM_006229477.2 XP_006229539.1 11, 12 1445

View Full Product Details