Gene Transcripts

Gene Symbol
RGD1563917
Entrez Gene ID
501176
Gene Name
similar to Nuclear autoantigen Sp-100 (Speckled 100 kDa)
Gene Aliases
Chromosome Location
Chr. 9: 92753841 - 92775830 on Build Rnor_6.0
UniGene ID
Rn.163087
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC088179.1 - 3 304

View Full Product Details