Gene Transcripts

Gene Symbol
Lrrc14b
Entrez Gene ID
502225
Gene Name
leucine rich repeat containing 14B
Gene Aliases
RGD1564978
Chromosome Location
Chr. 1: 31531143 - 31535314 on Build Rnor_6.0
UniGene ID
Rn.140570
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109333.1 NP_001102803.1 2 1309
GenBank mRNA FQ217859.1 - 2 1369

View Full Product Details