Gene Transcripts

Gene Symbol
Arf6
Entrez Gene ID
79121
Gene Name
ADP-ribosylation factor 6
Gene Aliases
Chromosome Location
Chr. 6: 91697109 - 91698257 on Build Rnor_6.0
UniGene ID
Rn.6993
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024152.2 NP_077066.1 1 487
GenBank mRNA BC091146.1 - 1 487
L12385.1 - 1 476

View Full Product Details