Gene Transcripts

Gene Symbol
Rab3b
Entrez Gene ID
81755
Gene Name
RAB3B, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 5: 128501787 - 128568170 on Build Rnor_6.0
UniGene ID
Rn.3788
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031091.2 NP_112353.1 4 544
GenBank mRNA BC060562.1 - 4 462
U68184.1 - 3 250
Y14019.1 - 3 432

View Full Product Details