Gene Transcripts

Gene Symbol
Timm8a1
Entrez Gene ID
84383
Gene Name
translocase of inner mitochondrial membrane 8 homolog A1 (yeast)
Gene Aliases
DDP, Ddp1, Timm8a
Chromosome Location
Chr. X: 105351714 - 105355722 on Build Rnor_6.0
UniGene ID
Rn.162342
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053370.3 NP_445822.1 292
GenBank mRNA BC060552.1 - 297

View Full Product Details