Gene Transcripts

Gene Symbol
CCL25
Entrez Gene ID
6370
Gene Name
C-C motif chemokine ligand 25
Gene Aliases
Ckb15, SCYA25, TECK
Chromosome Location
Chr.19: 8052318 - 8062663 on Build GRCh38
Uni Gene
Hs.310511
Interrogated Sequence Translated Protein
RefSeq NM_001201359.1 NP_001188288.1
NM_005624.3 NP_005615.2
XM_011528177.2 XP_011526479.1
XM_017027119.1 XP_016882608.1
XM_017027120.1 XP_016882609.1
XM_017027121.1 XP_016882610.1
XM_017027122.1 XP_016882611.1
GenBank mRNA AB046579.1 -
AI313430.1 -
AI961745.1 -
AK292662.1 -
BC125136.1 -
BC130561.1 -
BC144463.1 -
ENST00000390669.0 -
U86358.1 -

View Full Product Details