Gene Transcripts

Gene Symbol
CASP9
Entrez Gene ID
842
Gene Name
caspase 9
Gene Aliases
APAF-3, APAF3, ICE-LAP6, MCH6, PPP1R56
Chromosome Location
Chr.1: 15491401 - 15524912 on Build GRCh38
Uni Gene
Hs.329502
Interrogated Sequence Translated Protein
RefSeq NM_001229.4 NP_001220.2
NM_001278054.1 NP_001264983.1
NM_032996.3 NP_127463.2
NR_102732.1
NR_102733.1
XM_005246014.2 XP_005246071.1
XM_011542273.2 XP_011540575.1
GenBank mRNA AA534393.1 -
AB015653.1 -
AB020979.1 -
AB209147.1 -
AF093130.1 -
AF110376.1 -
AJ781267.1 -
AK292080.1 -
AK292111.1 -
AK303743.1 -
AY732490.2 -
BC002452.2 -
BC006463.1 -
BT006911.1 -
DA078665.1 -
ENST00000333868.0 -
HY027003.1 -
U56390.1 -
U60521.1 -

View Full Product Details