Gene Transcripts

Gene Symbol
FGFR3
Entrez Gene ID
2261
Gene Name
fibroblast growth factor receptor 3
Gene Aliases
ACH, CD333, CEK2, HSFGFR3EX, JTK4
Chromosome Location
Chr.4: 1793299 - 1808872 on Build GRCh38
Uni Gene
Hs.1420
Interrogated Sequence Translated Protein
RefSeq NM_000142.4 NP_000133.1
NM_001163213.1 NP_001156685.1
NM_022965.3 NP_075254.1
XM_006713868.1 XP_006713931.1
XM_006713869.1 XP_006713932.1
XM_006713870.1 XP_006713933.1
XM_006713871.1 XP_006713934.1
XM_006713872.1 XP_006713935.1
XM_006713873.1 XP_006713936.1
XM_011513420.1 XP_011511722.1
XM_011513422.1 XP_011511724.1
GenBank mRNA AB209441.1 -
AF238374.1 -
AF245114.1 -
AF369211.1 -
AF369212.1 -
AF369213.1 -
AK308936.1 -
AW204106.1 -
BC121175.2 -
BC128610.1 -
BC153824.1 -
DR003490.1 -
ENST00000440486.0 -
HM244683.1 -
KJ534834.1 -
KJ534974.1 -
KJ535004.1 -
M58051.1 -
M59374.1 -
M64347.1 -
X84939.1 -

View Full Product Details