Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001229.4 | NP_001220.2 |
NM_001278054.1 | NP_001264983.1 | |
NM_032996.3 | NP_127463.2 | |
NR_102732.1 | ||
NR_102733.1 | ||
XM_005246014.2 | XP_005246071.1 | |
XM_011542273.2 | XP_011540575.1 | |
GenBank mRNA | AA534393.1 | - |
AB015653.1 | - | |
AB020979.1 | - | |
AB209147.1 | - | |
AF093130.1 | - | |
AF110376.1 | - | |
AJ781267.1 | - | |
AK292080.1 | - | |
AK292111.1 | - | |
AK303743.1 | - | |
AY732490.2 | - | |
BC002452.2 | - | |
BC006463.1 | - | |
BT006911.1 | - | |
DA078665.1 | - | |
ENST00000333868.0 | - | |
HY027003.1 | - | |
U56390.1 | - | |
U60521.1 | - |