Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001201359.1 | NP_001188288.1 |
NM_005624.3 | NP_005615.2 | |
XM_011528177.2 | XP_011526479.1 | |
XM_017027119.1 | XP_016882608.1 | |
XM_017027120.1 | XP_016882609.1 | |
XM_017027121.1 | XP_016882610.1 | |
XM_017027122.1 | XP_016882611.1 | |
GenBank mRNA | AB046579.1 | - |
AI313430.1 | - | |
AI961745.1 | - | |
AK292662.1 | - | |
BC125136.1 | - | |
BC130561.1 | - | |
BC144463.1 | - | |
ENST00000390669.0 | - | |
U86358.1 | - |